Piperacillin sodium salt 835-1007 µg/mg
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Piperacillin sodium, Sodium (2S,5R,6R)-6-((R)-2-(4-ethyl-2,3-dioxopiperazine-1-carboxamido)-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
CAS No.: 59703-84-3
Molecular Formula: C23H26N5NaO7S
Formula Weight: 539.54
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to off-white
Odour: Odourless
Melting Point: 183-185°
Boiling Point: 793°
Solubility: Soluble in water or methanol
Beilstein Reference: 5373920
MDL No.: MFCD00917471
Applications: A broad spectrum antibiotic targeting a wide range of susceptible gram positive and gram negative bacteria
Formel:
C₂₃H₂₇N₅O₇S Molecular Weight: 539.54 g/mol Siedepunkt: 793 °C Schmelzpunkt: 183…185 °C |
MDL:
MFCD00056867 CAS-Nummer: 59703-84-3 EINECS: 261-868-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...