Apramycin sulphate
Lieferant: Thermo Fisher Scientific
Gefahr
Synonyme: (2R,3R,4S,5S,6S)-2-[[(2R,3S,4R,4aR,6S,7R,8aS)-7-amino-6-[(1R,2R,3S,4R,6S)-4,6-diamino-2,3-dihydroxycyclohexyl]oxy-4-hydroxy-3-(methylamino)-2,3,4,4a,6,7,8,8a-octahydropyrano[3,2-b]pyran-2-yl]oxy]-5-amino-6-(hydroxymethyl)oxane-3,4-diol, sulphuric acid, Nebramycin II, Apramycin sulphate salt, (2R,3R,4S,5S,6S)-5-Amino-2-(((2R,3S,4R,4aR,6S,7R,8aS)-7-amino-6-(((1R,2R,3S,4R,6S)-4,6-diamino-2,3-dihydroxycyclohexyl)oxy)-4-hydroxy-3-(methylamino)octahydropyrano[3,2-b]pyran-2-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4-diol xsulphate, Nebramycin II sulphate
J66616.03 J66616.09
J66616.03EA 62.5 EUR
J66616.03 J66616.09
Apramycin sulphate
Apramycin sulphate
Apramycin sulfate binds to the deep groove of RNA and effectively inhibits ribosomal translocation prohibiting protein synthesis.
  • Pale yellow powder
Soluble in water at 25mg/ml
Formel: C₂₁H₄₁N₅O₁₁·H₂SO₄
Molecular Weight: 637.65 g/mol
Siedepunkt: 823 °C
Schmelzpunkt: 168 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Kühlschrank
MDL: MFCD06200257
CAS-Nummer: 65710-07-8
EINECS: 265-890-7