Picloram 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Amino-3,5,6-trichloropicolinic Acid, 4-Amino-3,5,6-trichloropyridine-2-carboxylic Acid
Purity: 95%
CAS No.: 1918-02-1
Molecular Formula: C6H3Cl3N2O2
Formula Weight: 241.46
Storage Temperature: 2°C to 8°C
Flash Point:
Applications: Belongs to the puridine class of the auxinic compounds similar to the hormone Indole-3-acetic acid (IAA)
Formel:
C₆H₃Cl₃N₂O₂ Molecular Weight: 241,45 g/mol Siedepunkt: 421 °C Schmelzpunkt: 219 °C Flammpunkt: >110 °C (230 °F) |
MDL:
MFCD00012101 CAS-Nummer: 1918-02-1 EINECS: 217-636-1 Merck Index: 14,7397 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...