Sulphamonomethoxine 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Amino-N-(6-methoxypyrimidin-4-yl)benzenesulphonamide, 4-Amino-N-(6-methoxy-4-pyrimidinyl)benzenesulphonamide, N-(6-Methoxy-4-pyrimidinyl)sulphanilamide

J66733.MD J66733.03
J66733.MDEA 116 EUR
J66733.MD J66733.03
Sulphamonomethoxine 98%
Sulphamonomethoxine

Purity: 98% CAS No.: 1220-83-3 Molecular Formula: C11H12N4O3S Formula Weight: 280.30 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Physical Form: Powder Appearance: White to off-white Melting Point: 203-206° Boiling Point: 513° Solubility: Soluble in water at 10mg/ml with warming. Also soluble in ethanol or methanol Beilstein Reference: 621128 MDL No.: MFCD00063466


Applications: A bacteriostatic sulfonamide antibiotic which acts as a folic acid synthesis inhibitor

Formel: C₁₁H₁₂N₄O₃S
Molecular Weight: 280,30 g/mol
Siedepunkt: 513 °C
Schmelzpunkt: 203…206 °C
Flammpunkt: >110 °C (230 °F)
MDL: MFCD00063466
CAS-Nummer: 1220-83-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR