Florfenicol 98%
Lieferant: Thermo Scientific
Synonyme:
2,2-Dichloro-N-[(1R,2S)-3-fluoro-1-hydroxy-1-(4-methylsulphonylphenyl)-2-propyl]acetamide, D-(-)-threo-2-Dichloroacetamido-3-fluoro-1-(4-methylsulphonylphenyl)-1-propanol, 2,2-Dichloro-N-((1R,2S)-3-fluoro-1-hydroxy-1-(4-(methylsulphonyl)phenyl)propan-2-yl)acetamide
Purity: 98%
CAS No.: 73231-34-2
Molecular Formula: C12H14Cl2FNO4S
Formula Weight: 358.21
Flash Point:
Applications: A bacteriostatic antibiotic similar to chloramphenicol which inhibits bacterial protein synthesis
Formel:
C₁₂H₁₄Cl₂FNO₄S Molecular Weight: 358,21 g/mol Siedepunkt: 618 °C Schmelzpunkt: 152…156 °C Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00864834 CAS-Nummer: 73231-34-2 Merck Index: 14,4109 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...