Gold 99%, nanoparticles 100 nm reactant-free OD1 (Abs. 572 nm)
Lieferant: Thermo Fisher Scientific
Synonyme:
Aurum
J67004.AC
J67004.AE
J67004.ACEA
96.7
EUR
J67004.AC
J67004.AE
Gold 99%, nanoparticles 100 nm reactant-free OD1 (Abs. 572 nm)
Gold
Purity: 99%
CAS No.: 7440-57-5
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Pink to red
Odour: Odourless
MDL No.: MFCD00003436
Formel:
Au Molecular Weight: 196.97 g/mol Siedepunkt: 2800 Schmelzpunkt: 1063 Lagertemperatur: Kühlschrank |
MDL:
MFCD00003436 CAS-Nummer: 7440-57-5 Merck Index: 12,04526 |