1,9-Pyrazoloanthrone 99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
JNK Inhibitor II, Anthra[1,9-cd]pyrazol-6(2H)-one, Dibenzo[cd,g]indazol-6(2H)-one, SP600125, SP 600125
J67272.MC
J67272.MF
J67272.MCEA
182
EUR
J67272.MC
J67272.MF
1,9-Pyrazoloanthrone 99%
1,9-Pyrazoloanthrone
Purity: 99%
CAS No.: 129-56-6
Molecular Formula: C14H8N2O
Formula Weight: 220.23
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: Yellow to brown
Solubility: Soluble in DMSO at approx. 20mg/ml
MDL No.: MFCD00022289
Formel:
C₁₄H₈N₂O Molecular Weight: 220,231 g/mol |
MDL:
MFCD00022289 CAS-Nummer: 129-56-6 EINECS: 204-955-6 |