Gold 99%, nanoparticles 20 nm reactant-free OD1 (Abs. 524 nm)
Lieferant: Thermo Fisher Scientific
Synonyme: Aurum
J67370.AC J67370.AE
J67370.ACEA 102 EUR
J67370.AC J67370.AE
Gold 99%, nanoparticles 20 nm reactant-free OD1 (Abs. 524 nm)
Gold
Purity: 99% CAS No.: 7440-57-5 Storage Temperature: 2°C to 8°C Physical Form: Liquid Appearance: Pink to red Odour: Odourless MDL No.: MFCD00003436
Formel: Au
Molecular Weight: 196.97 g/mol
Siedepunkt: 2800
Schmelzpunkt: 1063
Lagertemperatur: Kühlschrank
MDL: MFCD00003436
CAS-Nummer: 7440-57-5
Merck Index: 12,04526