1,3-Diphenyl-2-benzofuran 97%

Lieferant: Thermo Scientific

Synonyme: 2,5-Diphenyl-3,4-benzofuran, 1,3-Diphenyl-isobenzofuran, DIB

L00101.03 L00101.06
L00101.03EA 33.2 EUR
L00101.03 L00101.06
1,3-Diphenyl-2-benzofuran 97%
1,3-Diphenylisobenzofuran

Purity: 97% CAS No.: 5471-63-6 Molecular Formula: C20H14O Formula Weight: 270.33 Storage Temperature: 2°C to 8°C Melting Point: 131-136° Beilstein Reference: 199922 MDL No.: MFCD00005931

Formel: C₂₀H₁₄O
Molecular Weight: 270,33 g/mol
Schmelzpunkt: 131…136 °C
MDL: MFCD00005931
CAS-Nummer: 5471-63-6
EINECS: 226-808-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR