Di-tert-butyl hydrazodicarboxylate ≥98%
Lieferant: Thermo Scientific
Synonyme: Di-tert-butyl hydrazine-1,2-dicarboxylate, Di-tert-butyl Hydrazodiformate, Di-tert-butyl 1,2-Hydrazinedicarboxylate
L00280.06 L00280.14
L00280.06EA 55.1 EUR
L00280.06 L00280.14
Di-tert-butyl hydrazodicarboxylate ≥98%
Di-tert-butyl hydrazodicarboxylate
Purity: 98+% CAS No.: 16466-61-8 Molecular Formula: C10H20N2O4 Formula Weight: 232.28 Melting Point: 123-126° Beilstein Reference: 1794023 MDL No.: MFCD00015000
Formel: C₁₀H₂₀N₂O₄
Molecular Weight: 232,28 g/mol
Schmelzpunkt: 123…126 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00015000
CAS-Nummer: 16466-61-8
EINECS: 240-512-3