N-tert-Butoxycarbonylimidazole 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: tert-Butyl 1H-imidazole-1-carboxylate, N-Boc-imidazole

L00506.06 L00506.14
L00506.06EA 26.1 EUR
L00506.06 L00506.14
N-tert-Butoxycarbonylimidazole 98%
N-tert-Butoxycarbonylimidazole

Purity: 98% CAS No.: 49761-82-2 Molecular Formula: C8H12N2O2 Formula Weight: 168.20 Storage Temperature: 2°C to 8°C Melting Point: 44-48° Beilstein Reference: 607792 MDL No.: MFCD00014497

Formel: C₈H₁₂N₂O₂
Molecular Weight: 168,20 g/mol
Schmelzpunkt: 44…48 °C
MDL: MFCD00014497
CAS-Nummer: 49761-82-2
EINECS: 256-475-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR