Diphenyl(Methyl)Sulfonium Tetrafluoroborat 95%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Methyldiphenylsulphonium Tetrafluoroborate, Diphenyl(methyl)sulphonium Tetrafluoroborate

L00512.06 L00512.14
L00512.06EA 82.7 EUR
L00512.06 L00512.14
Diphenyl(Methyl)Sulfonium Tetrafluoroborat 95%
Diphenyl(methyl)sulphonium tetrafluoroborate

Purity: 95% CAS No.: 10504-60-6 Molecular Formula: C13H13BF4S Formula Weight: 288.11 Melting Point: 63-67° Beilstein Reference: 4732019 UN No.: UN1759 MDL No.: MFCD00051861

Formel: C₁₃H₁₃BF₄S
Molecular Weight: 288.11 g/mol
Schmelzpunkt: 63…67 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00051861
CAS-Nummer: 10504-60-6
EINECS: 234-032-3
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR