5,6-Dimethoxy-1H-indole-2-carboxylic acid 98%
Lieferant: Thermo Scientific
Achtung
L00733.03 L00733.06
L00733.03EA 72.8 EUR
L00733.03 L00733.06
5,6-Dimethoxy-1H-indole-2-carboxylic acid 98%
5,6-Dimethoxy-1H-indole-2-carboxylic acid
Purity: 98% CAS No.: 88210-96-2 Molecular Formula: C11H11NO4 Formula Weight: 221.22 Melting Point: ca 210° dec. Beilstein Reference: 202249 MDL No.: MFCD00015459
Formel: C₁₁H₁₁NO₄
Molecular Weight: 221,21 g/mol
Schmelzpunkt: ∼ 210 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00015459
CAS-Nummer: 88210-96-2