Phenylpropiolaldehyde diethyl acetal 98%

Lieferant: Thermo Scientific

Synonyme: (3,3-Diethoxyprop-1-yn-1-yl)benzene

L00872.06 L00872.14
L00872.06EA 31.8 EUR
L00872.06 L00872.14
Phenylpropiolaldehyde diethyl acetal 98%
Phenylpropiolaldehyde diethyl acetal

Purity: 98% CAS No.: 6142-95-6 Molecular Formula: C6H5C≡CCH(OCH2CH3)2 Formula Weight: 204.27 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.5170 Boiling Point: 99-100°/2mm Beilstein Reference: 2049715 MDL No.: MFCD00009238

Formel: C₆H₅CΞCCH(OCH₂CH₃)₂
Molecular Weight: 204,27 g/mol
Siedepunkt: 99…100 °C (2 mmHg)
Dichte: 0,991
Flammpunkt: >110 °C (230 °F)
MDL: MFCD00009238
CAS-Nummer: 6142-95-6
EINECS: 228-143-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR