3-Brom-2,4,6-trimethylanilin ≥98%

Lieferant: Thermo Scientific
Gefahr

L00873.09 L00873.18
L00873.09EA 70.9 EUR
L00873.09 L00873.18
3-Brom-2,4,6-trimethylanilin ≥98%
3-Brom-2,4,6-trimethylanilin

Purity: 98+% CAS No.: 82842-52-2 Molecular Formula: C9H12BrN Formula Weight: 214.11 Flash Point: 110°(230°F) Melting Point: 34-38° Boiling Point: 153-155°/17mm Beilstein Reference: 2829829 MDL No.: MFCD00015476

Formel: C₉H₁₂BrN
Molecular Weight: 214,11 g/mol
Siedepunkt: 153…155 °C (17 mmHg)
Schmelzpunkt: 34…38 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00015476
CAS-Nummer: 82842-52-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR