5-Phenyl-2,4-pentadienoic acid ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
5-Phenylpenta-2,4-dienoic acid
L01004.06
L01004.14
L01004.06EA
76.8
EUR
L01004.06
L01004.14
5-Phenyl-2,4-pentadienoic acid ≥98%
5-Phenyl-2,4-pentadienoic acid
Purity: 98+%
CAS No.: 1552-94-9
Molecular Formula: C6H5CH=CHCH=CHCHCO2H
Formula Weight: 174.20
Melting Point: 163-167°
Beilstein Reference: 2042414
MDL No.: MFCD00014018
Formel:
C₆H₅CH=CHCH=CHCHCO₂H Molecular Weight: 174,20 g/mol Schmelzpunkt: 163…167 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014018 CAS-Nummer: 1552-94-9 EINECS: 216-298-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...