5-Benzyloxyindole-3-carbaldehyde 98%
Lieferant: Thermo Scientific
Synonyme:
5-(Benzyloxy)-1H-indole-3-carbaldehyde, 5-benzyloxy-1h-indole-3-carbaldehyde, 5-Benzyloxyindole-3-carboxaldehyde
L01117.03
L01117.06
L01117.03EA
60
EUR
L01117.03
L01117.06
5-Benzyloxyindole-3-carbaldehyde 98%
5-Benzyloxyindole-3-carbaldehyde
Purity: 98%
CAS No.: 6953-22-6
Molecular Formula: C16H13NO2
Formula Weight: 251.28
Melting Point: 239-242°
Beilstein Reference: 208873
MDL No.: MFCD00014562
Formel:
C₁₆H₁₃NO₂ Molecular Weight: 251,285 g/mol Schmelzpunkt: 239…242 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014562 CAS-Nummer: 6953-22-6 EINECS: 230-134-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...