2-Bromoacrylic acid 95%

Lieferant: Thermo Scientific
Gefahr

L01194.06 L01194.03 L01194.14
L01194.06EA 81.4 EUR
L01194.06 L01194.03 L01194.14
2-Bromoacrylic acid 95%
2-Bromoacrylic acid

Purity: 95% CAS No.: 10443-65-9 Molecular Formula: C3H3BrO2 Formula Weight: 150.96 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: Pale brown Melting Point: 65-70° Beilstein Reference: 1740582 UN No.: UN3261 MDL No.: MFCD00014333

Formel: C₃H₃BrO₂
Molecular Weight: 150,959 g/mol
Schmelzpunkt: 65…70 °C
MDL: MFCD00014333
CAS-Nummer: 10443-65-9
EINECS: 233-928-1
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR