3-Fluoro-DL-tyrosine 97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
H-DL-Tyr(3-F)-OH, 2-Amino-3-(3-fluoro-4-hydroxyphenyl)propanoic acid
L01479.MC
L01479.ME
L01479.MCEA
77.2
EUR
L01479.MC
L01479.ME
3-Fluoro-DL-tyrosine 97%
3-Fluoro-DL-tyrosine
Purity: 98%
CAS No.: 403-90-7
Molecular Formula: C9H10FNO3
Formula Weight: 199.18
Melting Point: ca 280° dec.
Beilstein Reference: 3204804
MDL No.: MFCD00063075
Formel:
C₉H₁₀FNO₃ Molecular Weight: 199,18 g/mol Schmelzpunkt: ∼ 280 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00063075 CAS-Nummer: 403-90-7 EINECS: 206-964-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...