D(-)-2-Phenylglycine ethyl ester hydrochloride ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
(R)-Ethyl 2-amino-2-phenylacetate hydrochloride, H-D-Phg-OEt.HCl, (R)-(-)-alpha-Aminophenylacetic acid ethyl ester hydrochloride
L01564.09
L01564.18
L01564.09EA
87.7
EUR
L01564.09
L01564.18
D(-)-2-Phenylglycine ethyl ester hydrochloride ≥98%
D(-)-2-Phenylglycine ethyl ester hydrochloride
Purity: 98+%
CAS No.: 17609-48-2
Molecular Formula: C10H13NO2.HCl
Formula Weight: 215.68
Melting Point: ca 190° dec.
Beilstein Reference: 5775455
Optical Rotation: -94° (c=1 in water)
MDL No.: MFCD00052005
Formel:
C₁₀H₁₃NO₂·HCl Schmelzpunkt: ∼ 190 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052005 CAS-Nummer: 17609-48-2 EINECS: 241-581-2 |