5,6-Dihydrouracil 97%
Lieferant: Thermo Scientific
Synonyme:
Dihydropyrimidine-2,4(1H,3H)-dione, Dihydrouracil
L01918.03
L01918.06
L01918.03EA
39.3
EUR
L01918.03
L01918.06
5,6-Dihydrouracil 97%
5,6-Dihydrouracil
Purity: 97%
CAS No.: 504-07-4
Molecular Formula: C4H6N2O2
Formula Weight: 114.10
Melting Point: 278-280°
Beilstein Reference: 112496
MDL No.: MFCD00006029
Formel:
C₄H₆N₂O₂ Molecular Weight: 114,10 g/mol Schmelzpunkt: 278…280 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006029 CAS-Nummer: 504-07-4 EINECS: 207-982-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...