5,6-Dihydro-5-methyluracil ≥98%

Lieferant: Thermo Scientific

Synonyme: 5-Methyldihydropyrimidine-2,4(1H,3H)-dione

L01996.14 L01996.06
L01996.14EA 130 EUR
L01996.14 L01996.06
5,6-Dihydro-5-methyluracil ≥98%
5,6-Dihydro-5-methyluracil

Purity: 98+% CAS No.: 696-04-8 Molecular Formula: C5H8N2O2 Formula Weight: 128.13 Melting Point: 263-265° Beilstein Reference: 81983 MDL No.: MFCD00023159

Formel: C₅H₈N₂O₂
Molecular Weight: 128,13 g/mol
Schmelzpunkt: 263…265 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00023159
CAS-Nummer: 696-04-8
EINECS: 211-787-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR