2-(p-Toluenesulphonyl)acetic acid 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Tosylacetic acid
L02102.06
L02102.14
L02102.06EA
98.6
EUR
L02102.06
L02102.14
2-(p-Toluenesulphonyl)acetic acid 98%
2-(p-Toluenesulphonyl)acetic acid
Purity: 98%
CAS No.: 3937-96-0
Molecular Formula: C9H10O4S
Formula Weight: 214.24
Melting Point: 117-119°
Beilstein Reference: 2331311
MDL No.: MFCD00021764
Formel:
C₉H₁₀O₄S Molecular Weight: 214.24 g/mol Schmelzpunkt: 117…119 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00021764 CAS-Nummer: 3937-96-0 EINECS: 223-518-0 |