Phenyl-p-tolylsulphone ≥98%
Lieferant: Thermo Scientific
Synonyme:
1-Methyl-4-(phenylsulphonyl)benzene
L03128.09
L03128.18
L03128.09EA
71.9
EUR
L03128.09
L03128.18
Phenyl-p-tolylsulphone ≥98%
Phenyl-p-tolylsulphone
Purity: 98+%
CAS No.: 640-57-3
Molecular Formula: C13H12O2S
Formula Weight: 232.30
Odour: Odourless
Melting Point: 125-129°
Beilstein Reference: 1872872
MDL No.: MFCD00025038
Formel:
C₁₃H₁₂O₂S Molecular Weight: 232,30 g/mol Schmelzpunkt: 125…129 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00025038 CAS-Nummer: 640-57-3 EINECS: 211-364-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...