4,4'-Biphenyldicarbonitril 98%

Lieferant: Thermo Fisher Scientific
Gefahr

L03450.03 L03450.06
L03450.03EA 77.6 EUR
L03450.03 L03450.06
4,4'-Biphenyldicarbonitril 98%
4,4'-Biphenyldicarbonitril

Purity: 98% CAS No.: 1591-30-6 Molecular Formula: C14H8N2 Formula Weight: 204.23 Melting Point: 232-236° Beilstein Reference: 1869917 UN No.: UN3439 MDL No.: MFCD00013805

Formel: C₁₄H₈N₂
Schmelzpunkt: 232…236 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013805
CAS-Nummer: 1591-30-6
EINECS: 216-468-6
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR