Phenylmaleic anhydride 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-Phenylfuran-2,5-dione, 3-Phenyl-2,5-furandione
L03453.03
L03453.06
L03453.03EA
26.5
EUR
L03453.03
L03453.06
Phenylmaleic anhydride 97%
Phenylmaleic anhydride
Purity: 97%
CAS No.: 36122-35-7
Molecular Formula: C10H6O3
Formula Weight: 174.16
Melting Point: 118-119°
Beilstein Reference: 127301
MDL No.: MFCD00015472
Formel:
C₁₀H₆O₃ Schmelzpunkt: 118…119 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00015472 CAS-Nummer: 36122-35-7 EINECS: 252-881-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...