2',3'-O-Isopropylideneuridine

Lieferant: Thermo Scientific

Synonyme: 1-((3AR,4R,6R,6aR)-6-(hydroxymethyl)-2,2-dimethyltetrahydrofuro[3,4-d][1,3]dioxol-4-yl)pyrimidine-2,4(1H,3H)-dione

L03686.03 L03686.06 L03686.14
L03686.03EA 47 EUR
L03686.03 L03686.06 L03686.14
2',3'-O-Isopropylideneuridine
2',3'-O-Isopropylideneuridine

CAS No.: 362-43-6 Molecular Formula: C12H16N2O6 Formula Weight: 284.27 Storage Temperature: 2°C to 8°C Melting Point: 165-166° Beilstein Reference: 32671 MDL No.: MFCD00034509

Formel: C₁₂H₁₆N₂O₆
Molecular Weight: 284,268 g/mol
Schmelzpunkt: 165…166 °C
MDL: MFCD00034509
CAS-Nummer: 362-43-6
EINECS: 206-647-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR