Ethyl-5-(benzyloxy)-1H-indole-2-carboxylate 97%

Lieferant: Thermo Scientific

Synonyme: 5-Benzyloxyindole-2-carboxylic acid ethyl ester, Ethyl 5-(benzyloxy)-1H-indole-2-carboxylate

L04440.03 L04440.06
L04440.03EA 41.4 EUR
L04440.03 L04440.06
Ethyl-5-(benzyloxy)-1H-indole-2-carboxylate 97%
Ethyl-5-(benzyloxy)-1H-indole-2-carboxylate

Purity: 97% CAS No.: 37033-95-7 Molecular Formula: C18H17NO3 Formula Weight: 295.34 Storage Temperature: -30°C to -10°C Melting Point: 160-165° Beilstein Reference: 281129 MDL No.: MFCD00022702

Formel: C₁₈H₁₇NO₃
Molecular Weight: 295,34 g/mol
Schmelzpunkt: 160…165 °C
MDL: MFCD00022702
CAS-Nummer: 37033-95-7
EINECS: 253-322-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR