DL-Mandelamide 97%
Lieferant: Thermo Scientific
Synonyme: 2-Hydroxy-2-phenylacetamide
L04679.03 L04679.06
L04679.03EA 42 EUR
L04679.03 L04679.06
DL-Mandelamide 97%
DL-Mandelamide
Purity: 97% CAS No.: 4410-31-5 Molecular Formula: C8H9NO2 Formula Weight: 151.16 Melting Point: 132-135° Beilstein Reference: 2208677 MDL No.: MFCD00025495
Formel: C₈H₉NO₂
Molecular Weight: 151,17 g/mol
Schmelzpunkt: 132…135 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00025495
CAS-Nummer: 4410-31-5