L-Adrenalin ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
L-Epinephrine
L04911.06
L04911.14
L04911.06EA
67.7
EUR
L04911.06
L04911.14
L-Adrenalin ≥98%
L-Adrenaline
Purity: 98+%
CAS No.: 51-43-4
Molecular Formula: C9H13NO3
Formula Weight: 183.20
Storage Temperature: 2°C to 8°C
Melting Point: 216-219°
Beilstein Reference: 2368277
Optical Rotation: -52° (c=1 in 0.6N HCl)
UN No.: UN2811
MDL No.: MFCD00002204
Formel:
C₉H₁₃NO₃ Molecular Weight: 183,21 g/mol Schmelzpunkt: 216…219 °C |
MDL:
MFCD00002204 CAS-Nummer: 51-43-4 EINECS: 200-098-7 ADR: 6.1,II Merck Index: 14,3619 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...