3-Bromo-2,4,6-trimethylnitrobenzene ≥90%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L05107.06
L05107.03
L05107.06EA
222
EUR
L05107.06
L05107.03
3-Bromo-2,4,6-trimethylnitrobenzene ≥90%
3-Bromo-2,4,6-trimethylnitrobenzene
Purity: 90+%
CAS No.: 90561-85-6
Molecular Formula: C9H10BrNO2
Formula Weight: 244.09
Melting Point: 56-62°
Boiling Point: 106-107°/1.3mm
Beilstein Reference: 1958703
MDL No.: MFCD00015924
Formel:
C₉H₁₀BrNO₂ Molecular Weight: 244.09 g/mol Siedepunkt: 106…107 °C (1.3 mmHg) Schmelzpunkt: 56…62 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00015924 CAS-Nummer: 90561-85-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...