3-[4-(Acetylamino)phenyl]acrylic acid überwiegend trans 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
trans-4-(Acetylamino)cinnamic acid
L05292.04
L05292.09
L05292.04EA
86.7
EUR
L05292.04
L05292.09
3-[4-(Acetylamino)phenyl]acrylic acid überwiegend trans 98%
3-[4-(Acetylamino)phenyl]acrylic acid
Purity: 98%
CAS No.: 7152-04-7
Molecular Formula: C11H11NO3
Formula Weight: 205.21
Melting Point: ca 259° dec.
Beilstein Reference: 2724869
MDL No.: MFCD00016846
Formel:
C₁₁H₁₁NO₃ Molecular Weight: 205.22 g/mol Schmelzpunkt: ∼ 259 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016846 CAS-Nummer: 7152-04-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...