(3-Cyanopropyl)triethoxysilane 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-(Triethoxysilyl)butanenitrile

L05367.14 L05367.22
L05367.14EA 48.6 EUR
L05367.14 L05367.22
(3-Cyanopropyl)triethoxysilane 97%
(3-Cyanopropyl)triethoxysilane

Purity: 97% CAS No.: 1067-47-6 Molecular Formula: C9H19NO3 Formula Weight: 231.37 Flash Point: 74°(165°F) Refractive Index: 1.4170 Boiling Point: 238-241° Beilstein Reference: 1942237 MDL No.: MFCD00042636

Formel: C₉H₁₉NO₃
Molecular Weight: 231,37 g/mol
Siedepunkt: 238…241 °C
Dichte: 0,961
Flammpunkt: 74 °C (165 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00042636
CAS-Nummer: 1067-47-6
EINECS: 213-931-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR