5-Hydroxy-3,4-dimethoxybenzoic acid ≥97%

Lieferant: Thermo Scientific
Achtung

L05708.03 L05708.MD
L05708.03EA 298 EUR
L05708.03 L05708.MD
5-Hydroxy-3,4-dimethoxybenzoic acid ≥97%
5-Hydroxy-3,4-dimethoxybenzoic acid

Purity: 97+% CAS No.: 1916-08-1 Molecular Formula: C9H10O5 Formula Weight: 198.17 Melting Point: 194-197° Beilstein Reference: 2115310 MDL No.: MFCD00002508

Formel: C₉H₁₀O₅
Molecular Weight: 198,17 g/mol
Schmelzpunkt: 194…197 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002508
CAS-Nummer: 1916-08-1
EINECS: 217-630-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR