2-Amino-4-imino-2-thiazoline hydrochloride 99%

Lieferant: Thermo Scientific
Gefahr

L05844.06 L05844.14
L05844.06EA 16.6 EUR
L05844.06 L05844.14
2-Amino-4-imino-2-thiazoline hydrochloride 99%
2-Amino-4-imino-2-thiazoline hydrochloride

Purity: 99% CAS No.: 36518-76-0 Molecular Formula: C3H5N3S.HCl Formula Weight: 151.62 Melting Point: 300° Beilstein Reference: 5109728 MDL No.: MFCD00012721

Formel: C₃H₅N₃S·HCl
Molecular Weight: 151,62 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012721
CAS-Nummer: 36518-76-0
EINECS: 253-083-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR