L-Arginin-methylester-dihydrochlorid 98%

Lieferant: Thermo Scientific

Synonyme: H-Arg-OMe·2HCL, H-Arg-OMe.2HCl

L05959.06 L05959.14
L05959.06EA 43.5 EUR
L05959.06 L05959.14
L-Arginin-methylester-dihydrochlorid 98%
L-Arginine methyl ester dihydrochloride

Purity: 98% CAS No.: 26340-89-6 Molecular Formula: C7H16N4O2.2HCl Formula Weight: 261.15 Melting Point: ca 190° dec. Beilstein Reference: 4159929 Optical Rotation: +21° (c=2.5 in methanol) MDL No.: MFCD00038948

Formel: C₇H₁₆N₄O₂
Molecular Weight: 261,15 g/mol
Schmelzpunkt: ∼ 190 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00038948
CAS-Nummer: 26340-89-6
EINECS: 247-622-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR