Dabsylchlorid ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: DABS-Cl, 4-Dimethylaminoazobenzene-4'-sulphonyl Chloride, 4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulphonyl chloride

L06057.MC L06057.ME
L06057.MCEA 28.8 EUR
L06057.MC L06057.ME
Dabsylchlorid ≥98%
Dabsyl chloride

Purity: 98+% CAS No.: 56512-49-3 Molecular Formula: C14H14ClN3O2S Formula Weight: 323.80 Storage Temperature: 2°C to 8°C Melting Point: ca 185° dec. Beilstein Reference: 3064095 UN No.: UN3261 MDL No.: MFCD00007444

Formel: C₁₄H₁₄ClN₃O₂S
Molecular Weight: 323,80 g/mol
Schmelzpunkt: ∼ 185 °C (Zers.)
MDL: MFCD00007444
CAS-Nummer: 56512-49-3
EINECS: 260-235-1
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR