2-(1H-Pyrrol-1-yl)benzoic acid 99%

Lieferant: Thermo Scientific
Achtung

L06069.03 L06069.06
L06069.03EA 44.3 EUR
L06069.03 L06069.06
2-(1H-Pyrrol-1-yl)benzoic acid 99%
2-(1H-Pyrrol-1-yl)benzoic acid

Purity: 99% CAS No.: 10333-68-3 Molecular Formula: C11H9NO2 Formula Weight: 187.20 Melting Point: 102-106° Beilstein Reference: 1343920 MDL No.: MFCD00051645

Formel: C₁₁H₉NO₂
Schmelzpunkt: 102…106 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00051645
CAS-Nummer: 10333-68-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR