Methyl-2-nitro-3,4,5-trimethoxybenzoate 98%

Lieferant: Thermo Fisher Scientific
Achtung

L06188.09 L06188.18
L06188.09EA 26.2 EUR
L06188.09 L06188.18
Methyl-2-nitro-3,4,5-trimethoxybenzoate 98%
Methyl-2-nitro-3,4,5-trimethoxybenzoate

Purity: 98% CAS No.: 5081-42-5 Molecular Formula: C11H13NO7 Formula Weight: 271.23 Melting Point: 66-70° Beilstein Reference: 2702653 MDL No.: MFCD00007097

Formel: C₁₁H₁₃NO₇
Molecular Weight: 271.23 g/mol
Schmelzpunkt: 66…70 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007097
CAS-Nummer: 5081-42-5
EINECS: 225-794-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR