Chlorcarbonyl-sulfenylchlorid 97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Chlorothioformyl Chloride, Chloroformylsulphenyl Chloride
L06432.03
L06432.06
L06432.03EA
24
EUR
L06432.03
L06432.06
Chlorcarbonyl-sulfenylchlorid 97%
Chlorocarbonylsulphenyl chloride
Purity: 97%
CAS No.: 2757-23-5
Molecular Formula: CCl2OS
Formula Weight: 130.98
Refractive Index: 1.5170
Boiling Point: 98-101°
Beilstein Reference: 506318
UN No.: UN3265
MDL No.: MFCD00000703
Formel:
CCl₂OS Molecular Weight: 130,97 g/mol Siedepunkt: 98…101 °C Dichte: 1,580 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000703 CAS-Nummer: 2757-23-5 EINECS: 220-415-2 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...