Ethyl-4-hydroxy-3-nitrobenzoate 98%

Lieferant: Thermo Scientific
Achtung

L06451.14 L06451.06
L06451.14EA 115 EUR
L06451.14 L06451.06
Ethyl-4-hydroxy-3-nitrobenzoate 98%
Ethyl-4-hydroxy-3-nitrobenzoate

Purity: 98% CAS No.: 19013-10-6 Molecular Formula: C9H9NO5 Formula Weight: 211.18 Melting Point: 69-71° Beilstein Reference: 2121319 MDL No.: MFCD00016999

Formel: C₉H₉NO₅
Molecular Weight: 211,18 g/mol
Schmelzpunkt: 69…71 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00016999
CAS-Nummer: 19013-10-6
EINECS: 242-751-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR