1-(2-Hydroxyethyl)hexamethylenimine 95%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2-(Azepan-1-yl)ethanol

L06497.06 L06497.14
L06497.06EA 25.3 EUR
L06497.06 L06497.14
1-(2-Hydroxyethyl)hexamethylenimine 95%
1-(2-Hydroxyethyl)hexamethylenimine

Purity: 95% CAS No.: 20603-00-3 Molecular Formula: C8H17NO Formula Weight: 143.22 Refractive Index: 1.4840 Boiling Point: 114-115°/23mm Beilstein Reference: 104110 UN No.: UN2735 MDL No.: MFCD00020988

Formel: C₈H₁₇NO
Molecular Weight: 143,23 g/mol
Siedepunkt: 114…115 °C (23 mmHg)
Dichte: 0,971
Lagertemperatur: Raumtemperatur
MDL: MFCD00020988
CAS-Nummer: 20603-00-3
EINECS: 243-915-2
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR