DL-Valinmethylesterhydrochlorid 99%

Lieferant: Thermo Scientific

Synonyme: H-DL-Val-OMe.HCl, H-DL-Val-OMe·HCl

L06526.06 L06526.14
L06526.06EA 43.9 EUR
L06526.06 L06526.14
DL-Valinmethylesterhydrochlorid 99%
DL-Valine methyl ester hydrochloride

Purity: 99% CAS No.: 5619-05-6 Molecular Formula: C6H13NO2.HCl Formula Weight: 167.64 Melting Point: 112-114° Beilstein Reference: 3912090 MDL No.: MFCD00066118

Formel: C₆H₁₃NO₂
Molecular Weight: 167,63 g/mol
Schmelzpunkt: 112…114 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00066118
CAS-Nummer: 5619-05-6
EINECS: 227-048-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR