Ethyl-2-amino-4-hydroxypyrimidine-5-carboxylate 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Ethyl 2-amino-4-hydroxypyrimidine-5-carboxylate
L06654.06
L06654.14
L06654.03
L06654.06EA
77.5
EUR
L06654.06
L06654.14
L06654.03
Ethyl-2-amino-4-hydroxypyrimidine-5-carboxylate 95%
Ethyl-2-amino-4-hydroxypyrimidine-5-carboxylate
Purity: 95%
CAS No.: 15400-53-0
Molecular Formula: C7H9N3O3
Formula Weight: 183.17
Odour: Odourless
Melting Point: ca 300° dec.
Beilstein Reference: 165835
MDL No.: MFCD00039708
Formel:
C₇H₉N₃O₃ Molecular Weight: 183.17 g/mol Schmelzpunkt: ∼ 300 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00039708 CAS-Nummer: 15400-53-0 EINECS: 239-418-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...