Nɑ-Acetyl-L-glutamine 99%
Lieferant: Thermo Scientific
Synonyme:
N-Acetyl-L-glutamine, N-Ac-Gln-OH, Ac-L-Gln-OH, Ac-Gln-OH
L06780.18
L06780.30
L06780.18EA
36.5
EUR
L06780.18
L06780.30
Nɑ-Acetyl-L-glutamine 99%
Nalpha-Acetyl-L-glutamine
Purity: 99%
CAS No.: 2490-97-3
Molecular Formula: C7H12N2O4
Formula Weight: 188.18
Storage Temperature: 2°C to 8°C
Melting Point: ca 206° dec.
Beilstein Reference: 1727471
Optical Rotation: -12° (c=3 in water)
MDL No.: MFCD00038159
Formel:
C₇H₁₂N₂O₄ Molecular Weight: 188,18 g/mol Schmelzpunkt: ∼ 206 °C (Zers.) |
MDL:
MFCD00038159 CAS-Nummer: 2490-97-3 EINECS: 219-647-7 Merck Index: 14,25 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...