Dimethylketene methyl trimethylsilyl acetal 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1-Methoxy-2-methyl-1-(trimethylsiloxy)propene, ((1-Methoxy-2-methylprop-1-en-1-yl)oxy)trimethylsilane

L06931.06 L06931.14
L06931.06EA 63.9 EUR
L06931.06 L06931.14
Dimethylketene methyl trimethylsilyl acetal 97%
Dimethylketene methyl trimethylsilyl acetal

Purity: 97% CAS No.: 31469-15-5 Molecular Formula: C8H18O2Si Formula Weight: 174.32 Flash Point: 30°(86°F) Refractive Index: 1.4150 Boiling Point: 35-36°/15mm Beilstein Reference: 1362893 UN No.: UN1993 MDL No.: MFCD00010232

Formel: C₈H₁₈O₂Si
Siedepunkt: 35…36 °C (15 mmHg)
Dichte: 0,858
Flammpunkt: 30 °C (86 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00010232
CAS-Nummer: 31469-15-5
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR