3,3-Dimethyl-1,2-epoxybutane 95%

Lieferant: Thermo Fisher Scientific
Gefahr

L06951.14 L06951.06
L06951.14EA 184 EUR
L06951.14 L06951.06
3,3-Dimethyl-1,2-epoxybutane 95%
3,3-Dimethyl-1,2-epoxybutane

Purity: 95% CAS No.: 2245-30-9 Molecular Formula: C6H12O Formula Weight: 100.16 Flash Point: -1°(30°F) Refractive Index: 1.4010 Odour: Recognizable Boiling Point: 95-96° Beilstein Reference: 102478 UN No.: UN1993 MDL No.: MFCD00051590

Formel: C₆H₁₂O
Siedepunkt: 95…96 °C
Dichte: 0,822
Flammpunkt: –1 °C (30 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00051590
CAS-Nummer: 2245-30-9
EINECS: 218-831-4
ADR: 3,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR