(Pyridin-2-ylsulphonyl)acetonitrile 98%

Lieferant: Thermo Scientific
Gefahr

L07114.03 L07114.06
L07114.03EA 77 EUR
L07114.03 L07114.06
(Pyridin-2-ylsulphonyl)acetonitrile 98%
(Pyridin-2-ylsulphonyl)acetonitrile

Purity: 98% CAS No.: 170449-34-0 Molecular Formula: C7H6N2O2S Formula Weight: 182.21 Melting Point: 77-79° UN No.: UN3439 MDL No.: MFCD00041443

Formel: C₇H₆N₂O₂S
Schmelzpunkt: 77…79 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00041443
CAS-Nummer: 170449-34-0
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR