4-Dimethylamino-1-naphthoic acid 98%
Lieferant: Thermo Scientific
|
Achtung
|
L07944.03
L07944.06
L07944.03EA
104
EUR
L07944.03
L07944.06
4-Dimethylamino-1-naphthoic acid 98%
4-Dimethylamino-1-naphthoic acid
Purity: 98%
CAS No.: 78062-03-0
Molecular Formula: C13H13NO2
Formula Weight: 215.25
Melting Point: ca 160° dec.
Beilstein Reference: 2723206
MDL No.: MFCD00051797
Formel:
C₁₃H₁₃NO₂ Molecular Weight: 215,25 g/mol Schmelzpunkt: ∼ 160 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051797 CAS-Nummer: 78062-03-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...