Methyl trans-2-hexenoate 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Methyl (E)-hex-2-enoate

L08030.09 L08030.18
L08030.09EA 53.4 EUR
L08030.09 L08030.18
Methyl trans-2-hexenoate 98%
Methyl trans-2-hexenoate

Purity: 98% CAS No.: 13894-63-8 Molecular Formula: C7H12O2 Formula Weight: 128.17 Flash Point: 45°(113°F) Refractive Index: 1.4345 Odour: Fruit-like Boiling Point: 57-58°/13mm Beilstein Reference: 1700768 UN No.: UN3272 MDL No.: MFCD00048798

Formel: C₇H₁₂O₂
Molecular Weight: 128,17 g/mol
Siedepunkt: 57…58 °C (13 mmHg)
Dichte: 0,915
Flammpunkt: 45 °C (113 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00048798
CAS-Nummer: 13894-63-8
EINECS: 237-663-2
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR